Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGTTACAGTGTCTGAAATTATA[A/C]CTGCAGACCAAGTCTTCTAGGCTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612014 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100506124 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100506124 - uncharacterized LOC100506124 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC21B - tetratricopeptide repeat domain 21B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024753.4 | 5163 | UTR 3 | NP_079029.3 | |||
XM_006712761.1 | 5163 | Intron | XP_006712824.1 | |||
XM_011511871.2 | 5163 | UTR 3 | XP_011510173.1 | |||
XM_011511872.2 | 5163 | UTR 3 | XP_011510174.1 | |||
XM_017004967.1 | 5163 | Intron | XP_016860456.1 | |||
XM_017004968.1 | 5163 | UTR 3 | XP_016860457.1 | |||
XM_017004969.1 | 5163 | UTR 3 | XP_016860458.1 |