Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606847 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCOF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TCOF1 - treacle ribosome biogenesis factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000356.3 | Intron | NP_000347.2 | ||||
NM_001008657.2 | Intron | NP_001008657.1 | ||||
NM_001135243.1 | Intron | NP_001128715.1 | ||||
NM_001135244.1 | Intron | NP_001128716.1 | ||||
NM_001135245.1 | Intron | NP_001128717.1 | ||||
NM_001195141.1 | Intron | NP_001182070.1 | ||||
XM_005268502.3 | Intron | XP_005268559.1 | ||||
XM_005268503.3 | Intron | XP_005268560.1 | ||||
XM_005268504.3 | Intron | XP_005268561.1 | ||||
XM_005268505.3 | Intron | XP_005268562.1 | ||||
XM_005268506.3 | Intron | XP_005268563.1 | ||||
XM_005268507.3 | Intron | XP_005268564.1 | ||||
XM_005268508.3 | Intron | XP_005268565.1 | ||||
XM_005268509.3 | Intron | XP_005268566.1 | ||||
XM_011537678.2 | Intron | XP_011535980.1 | ||||
XM_017009792.1 | Intron | XP_016865281.1 | ||||
XM_017009793.1 | Intron | XP_016865282.1 | ||||
XM_017009794.1 | Intron | XP_016865283.1 | ||||
XM_017009795.1 | Intron | XP_016865284.1 |
Set Membership: |
HapMap |