Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGATACCATTGATTTTTGCTTCC[A/G]CTGCAGGTTTATTATTATAATGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605409 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCERG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TCERG1 - transcription elongation regulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040006.1 | Intron | NP_001035095.1 | ||||
NM_006706.3 | Intron | NP_006697.2 | ||||
XM_005268365.2 | Intron | XP_005268422.1 | ||||
XM_017008977.1 | Intron | XP_016864466.1 | ||||
XM_017008978.1 | Intron | XP_016864467.1 | ||||
XM_017008979.1 | Intron | XP_016864468.1 | ||||
XM_017008980.1 | Intron | XP_016864469.1 | ||||
XM_017008981.1 | Intron | XP_016864470.1 | ||||
XM_017008982.1 | Intron | XP_016864471.1 | ||||
XM_017008983.1 | Intron | XP_016864472.1 | ||||
XM_017008984.1 | Intron | XP_016864473.1 | ||||
XM_017008985.1 | Intron | XP_016864474.1 |
Set Membership: |
HapMap |