Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACTTAGTCCATAAGTTCCTATTA[C/T]GCTTGCTTCTCTGAGTATATTTGGA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 605786 | |||||||||||||||||||||||
Literature Links: |
RBMS3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
RBMS3 - RNA binding motif single stranded interacting protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003792.2 | Intron | NP_001003792.1 | ||||
NM_001003793.2 | Intron | NP_001003793.1 | ||||
NM_001177711.1 | Intron | NP_001171182.1 | ||||
NM_001177712.1 | Intron | NP_001171183.1 | ||||
NM_014483.3 | Intron | NP_055298.2 | ||||
XM_005265060.2 | Intron | XP_005265117.1 | ||||
XM_005265061.2 | Intron | XP_005265118.1 | ||||
XM_005265062.1 | Intron | XP_005265119.1 | ||||
XM_005265063.1 | Intron | XP_005265120.1 | ||||
XM_005265065.4 | Intron | XP_005265122.1 | ||||
XM_017006178.1 | Intron | XP_016861667.1 | ||||
XM_017006179.1 | Intron | XP_016861668.1 | ||||
XM_017006180.1 | Intron | XP_016861669.1 | ||||
XM_017006181.1 | Intron | XP_016861670.1 | ||||
XM_017006182.1 | Intron | XP_016861671.1 |
RBMS3-AS3 - RBMS3 antisense RNA 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |