Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTCTCCCCTAGTCTCCACCCGCT[C/T]GACTCATGCCACCACCCTTGCCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612051 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BEAN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BEAN1 - brain expressed, associated with NEDD4, 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136106.4 | Intron | NP_001129578.1 | ||||
NM_001178020.2 | Intron | NP_001171491.1 | ||||
NM_001197224.3 | Intron | NP_001184153.1 | ||||
NM_001197225.3 | Intron | NP_001184154.1 | ||||
XM_011522883.1 | Intron | XP_011521185.1 | ||||
XM_011522884.1 | Intron | XP_011521186.1 | ||||
XM_011522885.2 | Intron | XP_011521187.1 | ||||
XM_011522887.2 | Intron | XP_011521189.1 | ||||
XM_011522888.1 | Intron | XP_011521190.1 | ||||
XM_011522889.1 | Intron | XP_011521191.1 | ||||
XM_011522890.1 | Intron | XP_011521192.1 | ||||
XM_011522891.1 | Intron | XP_011521193.1 | ||||
XM_011522892.2 | Intron | XP_011521194.1 | ||||
XM_011522894.2 | Intron | XP_011521196.1 | ||||
XM_011522895.1 | Intron | XP_011521197.1 |