Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTTCAAATAGGAATACTAGAGT[A/C]AGTGAGCTAGAAGAATTGAAGGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612425 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCTD18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCTD18 - potassium channel tetramerization domain containing 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SGO2 - shugoshin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160033.1 | Intron | NP_001153505.1 | ||||
NM_001160046.1 | Intron | NP_001153518.1 | ||||
NM_152524.5 | Intron | NP_689737.4 | ||||
XM_005246344.3 | Intron | XP_005246401.2 | ||||
XM_005246345.3 | Intron | XP_005246402.1 | ||||
XM_011510734.2 | Intron | XP_011509036.1 | ||||
XM_017003452.1 | Intron | XP_016858941.1 | ||||
XM_017003453.1 | Intron | XP_016858942.1 | ||||
XM_017003454.1 | Intron | XP_016858943.1 |