Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACAACAAAGGAACAGAACTAATA[G/T]TTGGGACCATAATCCAAGCATGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612425 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCTD18 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KCTD18 - potassium channel tetramerization domain containing 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SGO2 - shugoshin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160033.1 | Intron | NP_001153505.1 | ||||
NM_001160046.1 | Intron | NP_001153518.1 | ||||
NM_152524.5 | Intron | NP_689737.4 | ||||
XM_005246344.3 | Intron | XP_005246401.2 | ||||
XM_005246345.3 | Intron | XP_005246402.1 | ||||
XM_011510734.2 | Intron | XP_011509036.1 | ||||
XM_017003452.1 | Intron | XP_016858941.1 | ||||
XM_017003453.1 | Intron | XP_016858942.1 | ||||
XM_017003454.1 | Intron | XP_016858943.1 |
Set Membership: |
HapMap |