Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602633 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf49 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
C2orf49 - chromosome 2 open reading frame 49 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FHL2 - four and a half LIM domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039492.2 | Intron | NP_001034581.1 | ||||
NM_001318894.1 | Intron | NP_001305823.1 | ||||
NM_001318895.1 | Intron | NP_001305824.1 | ||||
NM_001318896.1 | Intron | NP_001305825.1 | ||||
NM_001318897.1 | Intron | NP_001305826.1 | ||||
NM_001318898.1 | Intron | NP_001305827.1 | ||||
NM_001318899.1 | Intron | NP_001305828.1 | ||||
NM_001450.3 | Intron | NP_001441.4 | ||||
NM_201555.1 | Intron | NP_963849.1 | ||||
NM_201557.3 | Intron | NP_963851.2 | ||||
XM_011510798.2 | Intron | XP_011509100.1 |
Set Membership: |
HapMap Validated |