Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCCAGCTGTATGGTAATGGGCG[A/G]CGCGGCCACTCTGCTTAACTCGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602576 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LFNG PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LFNG - LFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040167.1 | Intron | NP_001035257.1 | ||||
NM_001040168.1 | Intron | NP_001035258.1 | ||||
NM_001166355.1 | Intron | NP_001159827.1 | ||||
NM_002304.2 | Intron | NP_002295.1 |
MIR4648 - microRNA 4648 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |