Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTGGACACGCTTTAGATTCAAT[A/G]GGAAGAAAAGGATTTTTTCTTCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616404 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC90784 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC90784 - uncharacterized LOC90784 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR1A - RNA polymerase I subunit A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015425.4 | Intron | NP_056240.2 |
Set Membership: |
HapMap Validated |