Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605939 MIM: 604083 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLCD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PLCD4 - phospholipase C delta 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF142 - zinc finger protein 142 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105537.2 | 6221 | Silent Mutation | GAA,GAG | E,E 1671 | NP_001099007.1 | |
XM_006712730.3 | 6221 | Silent Mutation | GAA,GAG | E,E 1871 | XP_006712793.1 | |
XM_011511784.2 | 6221 | Silent Mutation | GAA,GAG | E,E 1871 | XP_011510086.1 | |
XM_011511785.2 | 6221 | Silent Mutation | GAA,GAG | E,E 1871 | XP_011510087.1 | |
XM_011511786.2 | 6221 | Silent Mutation | GAA,GAG | E,E 1871 | XP_011510088.1 | |
XM_011511787.2 | 6221 | Silent Mutation | GAA,GAG | E,E 1871 | XP_011510089.1 | |
XM_011511788.2 | 6221 | Silent Mutation | GAA,GAG | E,E 1871 | XP_011510090.1 | |
XM_011511789.2 | 6221 | Silent Mutation | GAA,GAG | E,E 1671 | XP_011510091.1 | |
XM_017004871.1 | 6221 | Silent Mutation | GAA,GAG | E,E 1871 | XP_016860360.1 | |
XM_017004872.1 | 6221 | Silent Mutation | GAA,GAG | E,E 1671 | XP_016860361.1 |
Set Membership: |
HapMap |