Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGTGCCAGTGGGAGCAGTGACAG[C/T]GAGGGGTGGTGGCCCTGCGGCCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602214 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSNK1G2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSNK1G2 - casein kinase 1 gamma 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319.6 | Intron | NP_001310.3 | ||||
XM_005259498.1 | Intron | XP_005259555.1 | ||||
XM_005259499.3 | Intron | XP_005259556.1 | ||||
XM_005259500.1 | Intron | XP_005259557.1 | ||||
XM_005259501.2 | Intron | XP_005259558.1 | ||||
XM_017026297.1 | Intron | XP_016881786.1 | ||||
XM_017026298.1 | Intron | XP_016881787.1 |
CSNK1G2-AS1 - CSNK1G2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |