Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAAACTTTCCACTGCTTTCTGAG[C/T]TTTAGTAAGATTTGGAGGCATATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604045 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101926933 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC101926933 - uncharacterized LOC101926933 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRMT5 - protein arginine methyltransferase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039619.2 | Intron | NP_001034708.1 | ||||
NM_001282953.1 | Intron | NP_001269882.1 | ||||
NM_001282954.1 | Intron | NP_001269883.1 | ||||
NM_001282955.1 | Intron | NP_001269884.1 | ||||
NM_001282956.1 | Intron | NP_001269885.1 | ||||
NM_006109.4 | Intron | NP_006100.2 | ||||
XM_017020927.1 | Intron | XP_016876416.1 |
PRMT5-AS1 - PRMT5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBM23 - RNA binding motif protein 23 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |