Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP221 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CFAP221 - cilia and flagella associated protein 221 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271049.1 | Intron | NP_001257978.1 | ||||
XM_006712353.3 | Intron | XP_006712416.1 | ||||
XM_017003550.1 | Intron | XP_016859039.1 | ||||
XM_017003551.1 | Intron | XP_016859040.1 | ||||
XM_017003552.1 | Intron | XP_016859041.1 | ||||
XM_017003553.1 | Intron | XP_016859042.1 | ||||
XM_017003554.1 | Intron | XP_016859043.1 | ||||
XM_017003555.1 | Intron | XP_016859044.1 | ||||
XM_017003556.1 | Intron | XP_016859045.1 | ||||
XM_017003557.1 | Intron | XP_016859046.1 | ||||
XM_017003558.1 | Intron | XP_016859047.1 | ||||
XM_017003559.1 | Intron | XP_016859048.1 |
Set Membership: |
Validated |