Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCCGCCTGAGCTTGGAGCTTGGC[A/G]TGGAGGAGTGCTGCTTCGGTCTACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 110750 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GYPC PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GYPC - glycophorin C (Gerbich blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256584.1 | Intron | NP_001243513.1 | ||||
NM_002101.4 | Intron | NP_002092.1 | ||||
NM_016815.3 | Intron | NP_058131.1 | ||||
XM_006712460.3 | Intron | XP_006712523.1 |