Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602119 | ||||||||||||||||||||
Literature Links: |
CHD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHD2 - chromodomain helicase DNA binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042572.2 | Intron | NP_001036037.1 | ||||
NM_001271.3 | Intron | NP_001262.3 |
LINC01578 - long intergenic non-protein coding RNA 1578 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3175 - microRNA 3175 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |