Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTTTCAGTGTAAGTTCGAGCCTT[A/G]GGAGGGTTGGGGACCCCAGATGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604883 MIM: 137960 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTF3C2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GTF3C2 - general transcription factor IIIC subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035521.2 | 3089 | Silent Mutation | CCC,CCT | P,P 782 | NP_001030598.1 | |
NM_001318909.1 | 3089 | Silent Mutation | CCC,CCT | P,P 793 | NP_001305838.1 | |
NM_001521.3 | 3089 | Silent Mutation | CCC,CCT | P,P 782 | NP_001512.1 | |
XM_005264272.4 | 3089 | Silent Mutation | CCC,CCT | P,P 821 | XP_005264329.1 | |
XM_011532801.2 | 3089 | Silent Mutation | CCC,CCT | P,P 897 | XP_011531103.1 |
GTF3C2-AS1 - GTF3C2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MPV17 - MPV17, mitochondrial inner membrane protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |