Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614241 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LCLAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LCLAT1 - lysocardiolipin acyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002257.2 | Intron | NP_001002257.1 | ||||
NM_001304445.1 | Intron | NP_001291374.1 | ||||
NM_001304446.1 | Intron | NP_001291375.1 | ||||
NM_182551.4 | Intron | NP_872357.2 | ||||
XM_005264244.1 | Intron | XP_005264301.1 | ||||
XM_005264245.3 | Intron | XP_005264302.1 | ||||
XM_011532741.2 | Intron | XP_011531043.1 | ||||
XM_011532742.2 | Intron | XP_011531044.1 | ||||
XM_011532743.2 | Intron | XP_011531045.1 | ||||
XM_011532744.2 | Intron | XP_011531046.1 | ||||
XM_011532745.1 | Intron | XP_011531047.1 | ||||
XM_017003746.1 | Intron | XP_016859235.1 | ||||
XM_017003747.1 | Intron | XP_016859236.1 |
Set Membership: |
HapMap |