Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607380 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAF3IP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TRAF3IP1 - TRAF3 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139490.1 | Intron | NP_001132962.1 | ||||
NM_015650.3 | Intron | NP_056465.2 | ||||
XM_006712414.2 | Intron | XP_006712477.1 | ||||
XM_011510944.2 | Intron | XP_011509246.1 | ||||
XM_011510945.2 | Intron | XP_011509247.1 | ||||
XM_011510946.2 | Intron | XP_011509248.1 | ||||
XM_011510947.2 | Intron | XP_011509249.1 | ||||
XM_011510948.2 | Intron | XP_011509250.1 | ||||
XM_011510950.2 | Intron | XP_011509252.1 | ||||
XM_017003789.1 | Intron | XP_016859278.1 |