Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615748 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1033 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
|||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
KIAA1033 - KIAA1033 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293640.1 | Intron | NP_001280569.1 | ||||
NM_015275.2 | Intron | NP_056090.1 | ||||
XM_011538073.2 | Intron | XP_011536375.1 | ||||
XM_011538074.2 | Intron | XP_011536376.1 | ||||
XM_011538077.2 | Intron | XP_011536379.1 | ||||
XM_017019082.1 | Intron | XP_016874571.1 |
LOC414300 - uncharacterized LOC414300 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |