Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608490 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC38A1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC38A1 - solute carrier family 38 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077484.1 | 7093 | UTR 3 | NP_001070952.1 | |||
NM_001278387.1 | 7093 | UTR 3 | NP_001265316.1 | |||
NM_001278388.1 | 7093 | UTR 3 | NP_001265317.1 | |||
NM_001278389.1 | 7093 | UTR 3 | NP_001265318.1 | |||
NM_001278390.1 | 7093 | Intron | NP_001265319.1 | |||
NM_030674.3 | 7093 | UTR 3 | NP_109599.3 | |||
XM_011538784.2 | 7093 | Intron | XP_011537086.1 | |||
XM_011538786.2 | 7093 | Intron | XP_011537088.1 | |||
XM_011538787.2 | 7093 | Intron | XP_011537089.1 | |||
XM_017019990.1 | 7093 | Intron | XP_016875479.1 |
Set Membership: |
HapMap |