Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCTCACCAGGCCCCAGGCCGTCT[A/G]ATCTGTCTGCTGCCTTGTTCCCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600950 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AANAT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AANAT - aralkylamine N-acetyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001088.2 | Intron | NP_001079.1 | ||||
NM_001166579.1 | Intron | NP_001160051.1 | ||||
XM_017024259.1 | Intron | XP_016879748.1 |
UBE2O - ubiquitin conjugating enzyme E2 O | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |