Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTCTACCAGTCCCTGATAACTA[C/T]TCAAGCTCCCTTTAAAGAGGCTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603312 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BBOX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
BBOX1 - gamma-butyrobetaine hydroxylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003986.2 | 87 | Intron | NP_003977.1 | |||
XM_005253159.2 | 87 | UTR 5 | XP_005253216.1 | |||
XM_005253160.2 | 87 | UTR 5 | XP_005253217.1 | |||
XM_005253161.2 | 87 | Intron | XP_005253218.1 | |||
XM_011520402.1 | 87 | UTR 5 | XP_011518704.1 | |||
XM_017018403.1 | 87 | Intron | XP_016873892.1 |
BBOX1-AS1 - BBOX1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |