Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607126 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLPP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PLPP2 - phospholipid phosphatase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003712.3 | 638 | Missense Mutation | GAG,GGG | E,G 180 | NP_003703.1 | |
NM_177526.2 | 638 | Missense Mutation | GAG,GGG | E,G 124 | NP_803545.1 | |
NM_177543.2 | 638 | Missense Mutation | GAG,GGG | E,G 201 | NP_808211.1 | |
XM_011528396.2 | 638 | Missense Mutation | GAG,GGG | E,G 186 | XP_011526698.1 |
Set Membership: |
HapMap |