Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTGTTTGCCCTTGAAGGCTTTC[A/G]CCGATTTCTAGAGATAAATCAAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602167 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESRRB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ESRRB - estrogen related receptor beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004452.3 | Intron | NP_004443.3 | ||||
XM_011536547.2 | Intron | XP_011534849.1 | ||||
XM_011536550.2 | Intron | XP_011534852.1 | ||||
XM_011536553.2 | Intron | XP_011534855.1 | ||||
XM_011536554.2 | Intron | XP_011534856.1 | ||||
XM_017021085.1 | Intron | XP_016876574.1 | ||||
XM_017021086.1 | Intron | XP_016876575.1 |