Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGAACACCACACTACTACGTTTT[C/T]AAGCTGTAATGGGAAGGAAAACACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607648 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STK39 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
STK39 - serine/threonine kinase 39 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013233.2 | 2446 | Intron | NP_037365.2 | |||
XM_005246465.2 | 2446 | Intron | XP_005246522.1 | |||
XM_017003813.1 | 2446 | UTR 3 | XP_016859302.1 | |||
XM_017003814.1 | 2446 | Intron | XP_016859303.1 | |||
XM_017003815.1 | 2446 | Intron | XP_016859304.1 | |||
XM_017003816.1 | 2446 | Intron | XP_016859305.1 | |||
XM_017003817.1 | 2446 | Intron | XP_016859306.1 |