Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601254 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP2K6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAP2K6 - mitogen-activated protein kinase kinase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002758.3 | Intron | NP_002749.2 | ||||
XM_005257515.2 | Intron | XP_005257572.1 | ||||
XM_005257516.2 | Intron | XP_005257573.1 | ||||
XM_006721975.3 | Intron | XP_006722038.1 | ||||
XM_011525025.2 | Intron | XP_011523327.1 | ||||
XM_011525026.2 | Intron | XP_011523328.1 | ||||
XM_011525027.2 | Intron | XP_011523329.1 |