Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601060 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENPP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ENPP2 - ectonucleotide pyrophosphatase/phosphodiesterase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040092.2 | 2562 | Silent Mutation | AGG,CGG | R,R 826 | NP_001035181.1 | |
NM_001130863.2 | 2562 | Silent Mutation | AGG,CGG | R,R 851 | NP_001124335.1 | |
NM_006209.4 | 2562 | Silent Mutation | AGG,CGG | R,R 878 | NP_006200.3 | |
XM_006716584.1 | 2562 | Silent Mutation | AGG,CGG | R,R 903 | XP_006716647.1 | |
XM_006716585.1 | 2562 | Silent Mutation | AGG,CGG | R,R 899 | XP_006716648.1 | |
XM_006716587.1 | 2562 | Silent Mutation | AGG,CGG | R,R 874 | XP_006716650.1 | |
XM_017013571.1 | 2562 | Silent Mutation | AGG,CGG | R,R 847 | XP_016869060.1 | |
XM_017013572.1 | 2562 | Silent Mutation | AGG,CGG | R,R 847 | XP_016869061.1 | |
XM_017013573.1 | 2562 | Silent Mutation | AGG,CGG | R,R 843 | XP_016869062.1 | |
XM_017013574.1 | 2562 | Silent Mutation | AGG,CGG | R,R 822 | XP_016869063.1 | |
XM_017013575.1 | 2562 | Silent Mutation | AGG,CGG | R,R 822 | XP_016869064.1 |