Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606733 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OSBPL5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
|||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
OSBPL5 - oxysterol binding protein like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144063.1 | 4074 | UTR 3 | NP_001137535.1 | |||
NM_020896.3 | 4074 | UTR 3 | NP_065947.1 | |||
NM_145638.2 | 4074 | UTR 3 | NP_663613.1 | |||
XM_011519873.2 | 4074 | UTR 3 | XP_011518175.1 | |||
XM_017017163.1 | 4074 | UTR 3 | XP_016872652.1 | |||
XM_017017164.1 | 4074 | UTR 3 | XP_016872653.1 |