Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAAAGGTCCTGCTTCTAAATGCA[G/T]TCACATTCTAAGGTCCTGGGGGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612299 MIM: 611728 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COMMD9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COMMD9 - COMM domain containing 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRR5L - proline rich 5 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160167.1 | Intron | NP_001153639.1 | ||||
NM_001160168.1 | Intron | NP_001153640.1 | ||||
NM_001160169.1 | Intron | NP_001153641.1 | ||||
NM_024841.4 | Intron | NP_079117.3 |