Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf74 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
C2orf74 - chromosome 2 open reading frame 74 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143959.1 | 96 | Intron | NP_001137431.1 | |||
NM_001143960.2 | 96 | UTR 5 | NP_001137432.1 | |||
NM_001316317.1 | 96 | UTR 5 | NP_001303246.1 | |||
XM_017003967.1 | 96 | Intron | XP_016859456.1 |
KIAA1841 - KIAA1841 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005264605.3 | 96 | Intron | XP_005264662.1 | |||
XM_005264606.4 | 96 | Intron | XP_005264663.1 | |||
XM_005264607.2 | 96 | Intron | XP_005264664.1 | |||
XM_011533126.2 | 96 | Intron | XP_011531428.1 | |||
XM_011533127.2 | 96 | Intron | XP_011531429.1 | |||
XM_011533128.1 | 96 | Intron | XP_011531430.1 | |||
XM_011533129.2 | 96 | Intron | XP_011531431.1 | |||
XM_011533131.2 | 96 | Intron | XP_011531433.1 | |||
XM_017005108.1 | 96 | Intron | XP_016860597.1 | |||
XM_017005109.1 | 96 | Intron | XP_016860598.1 |
LOC339803 - uncharacterized LOC339803 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |