Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608148 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SATB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SATB2 - SATB homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172509.1 | Intron | NP_001165980.1 | ||||
NM_001172517.1 | Intron | NP_001165988.1 | ||||
NM_015265.3 | Intron | NP_056080.1 | ||||
XM_005246396.2 | Intron | XP_005246453.1 | ||||
XM_006712372.2 | Intron | XP_006712435.1 | ||||
XM_011510840.2 | Intron | XP_011509142.1 | ||||
XM_017003656.1 | Intron | XP_016859145.1 |
Set Membership: |
HapMap Validated |