Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATCTTTCCTTTGGAAGCAGTAT[A/C]CATAAGCCTTCATCAGAAGCTTCTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602904 | ||||||||||||||||||||
Literature Links: |
MAPK6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAPK6 - mitogen-activated protein kinase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002748.3 | 39 | Intron | NP_002739.1 | |||
XM_005254537.2 | 39 | Intron | XP_005254594.1 | |||
XM_005254538.2 | 39 | Intron | XP_005254595.1 | |||
XM_005254539.3 | 39 | UTR 5 | XP_005254596.1 | |||
XM_011521782.1 | 39 | Intron | XP_011520084.1 |