Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGCCTAGGGGTTGGTGATTCTTA[A/C]GTTGTTGTTCCACAGGTTACAGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614248 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PEAK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PEAK1 - pseudopodium enriched atypical kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024776.3 | 8862 | UTR 3 | NP_079052.2 | |||
XM_005254670.2 | 8862 | UTR 3 | XP_005254727.1 | |||
XM_005254671.2 | 8862 | UTR 3 | XP_005254728.1 | |||
XM_005254673.2 | 8862 | UTR 3 | XP_005254730.1 | |||
XM_005254674.2 | 8862 | UTR 3 | XP_005254731.1 | |||
XM_005254675.4 | 8862 | UTR 3 | XP_005254732.1 | |||
XM_011522033.2 | 8862 | UTR 3 | XP_011520335.1 | |||
XM_011522034.2 | 8862 | UTR 3 | XP_011520336.1 | |||
XM_011522035.2 | 8862 | UTR 3 | XP_011520337.1 | |||
XM_011522036.2 | 8862 | UTR 3 | XP_011520338.1 | |||
XM_011522037.2 | 8862 | UTR 3 | XP_011520339.1 | |||
XM_011522038.2 | 8862 | UTR 3 | XP_011520340.1 | |||
XM_011522039.2 | 8862 | UTR 3 | XP_011520341.1 | |||
XM_011522040.1 | 8862 | Intron | XP_011520342.1 | |||
XM_011522041.1 | 8862 | Intron | XP_011520343.1 | |||
XM_011522042.2 | 8862 | UTR 3 | XP_011520344.1 |
Set Membership: |
HapMap Validated |