Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTCCTTGTTCAAAAATGGAAAT[C/T]GAATTTGTAAAGACTAACAAGAGGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606594 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SETD7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SETD7 - SET domain containing lysine methyltransferase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306199.1 | Intron | NP_001293128.1 | ||||
NM_001306200.1 | Intron | NP_001293129.1 | ||||
NM_030648.3 | Intron | NP_085151.1 | ||||
XM_017008661.1 | Intron | XP_016864150.1 |
Set Membership: |
HapMap |