Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATAAATTTGAAATAATATCCACAT[C/G]ACAGTAATTGTGATAATGAAAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607156 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ST3GAL6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ST3GAL6 - ST3 beta-galactoside alpha-2,3-sialyltransferase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271142.1 | Intron | NP_001258071.1 | ||||
NM_001271145.1 | Intron | NP_001258074.1 | ||||
NM_001271146.1 | Intron | NP_001258075.1 | ||||
NM_001271147.1 | Intron | NP_001258076.1 | ||||
NM_001271148.1 | Intron | NP_001258077.1 | ||||
NM_001323352.1 | Intron | NP_001310281.1 | ||||
NM_001323353.1 | Intron | NP_001310282.1 | ||||
NM_001323358.1 | Intron | NP_001310287.1 | ||||
NM_001323359.1 | Intron | NP_001310288.1 | ||||
NM_001323360.1 | Intron | NP_001310289.1 | ||||
NM_001323362.1 | Intron | NP_001310291.1 | ||||
NM_001323363.1 | Intron | NP_001310292.1 | ||||
NM_001323364.1 | Intron | NP_001310293.1 | ||||
NM_001323365.1 | Intron | NP_001310294.1 | ||||
NM_001323366.1 | Intron | NP_001310295.1 | ||||
NM_001323367.1 | Intron | NP_001310296.1 | ||||
NM_001323368.1 | Intron | NP_001310297.1 | ||||
NM_006100.3 | Intron | NP_006091.1 |
ST3GAL6-AS1 - ST3GAL6 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |