Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAATACAAACATGCTACAGTTACC[C/T]TGTCTATCCGAACTGTTAAGCCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600025 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KLC1 - kinesin light chain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130107.1 | Intron | NP_001123579.1 | ||||
NM_005552.4 | Intron | NP_005543.2 | ||||
NM_182923.3 | Intron | NP_891553.2 |
Set Membership: |
HapMap |