Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTTGTTCATAACTATGTATAAA[C/T]AGTTTTCTATCACTATCACTGCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602385 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SULT1C2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SULT1C2 - sulfotransferase family 1C member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001056.3 | Intron | NP_001047.1 | ||||
NM_176825.2 | Intron | NP_789795.1 | ||||
XM_005264013.3 | Intron | XP_005264070.1 |
Set Membership: |
HapMap |