Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614693 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATMIN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATMIN - ATM interactor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C16orf46 - chromosome 16 open reading frame 46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100873.1 | Intron | NP_001094343.1 | ||||
NM_152337.2 | Intron | NP_689550.2 | ||||
XM_005255792.2 | Intron | XP_005255849.1 | ||||
XM_011522848.2 | Intron | XP_011521150.2 | ||||
XM_011522849.2 | Intron | XP_011521151.2 | ||||
XM_017022916.1 | Intron | XP_016878405.1 | ||||
XM_017022917.1 | Intron | XP_016878406.1 |