Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATGCTGGTTGTGACCTGGATTA[A/G]TTTCACAACCTACAAAATGTTCGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100506302 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC100506302 - uncharacterized LOC100506302 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBM33 - RNA binding motif protein 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_053043.2 | Intron | NP_444271.2 | ||||
XM_005249531.1 | Intron | XP_005249588.1 | ||||
XM_005249532.1 | Intron | XP_005249589.1 | ||||
XM_005249533.1 | Intron | XP_005249590.1 | ||||
XM_005249534.1 | Intron | XP_005249591.1 | ||||
XM_005249535.1 | Intron | XP_005249592.1 | ||||
XM_011515840.1 | Intron | XP_011514142.1 |