Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615380 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AREL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AREL1 - apoptosis resistant E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039479.1 | Intron | NP_001034568.1 | ||||
XM_005268244.3 | Intron | XP_005268301.1 | ||||
XM_006720344.2 | Intron | XP_006720407.1 | ||||
XM_011537415.2 | Intron | XP_011535717.1 | ||||
XM_011537416.2 | Intron | XP_011535718.1 | ||||
XM_011537417.2 | Intron | XP_011535719.1 |
Set Membership: |
HapMap |