Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCATCCTCCTCCTCTTCCTCGCA[A/G]TCCTATTTGAGCCATTCCCAGTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
YLPM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
YLPM1 - YLP motif containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019589.2 | 813 | Silent Mutation | CAA,CAG | Q,Q 215 | NP_062535.2 | |
XM_005267860.3 | 813 | Silent Mutation | CAA,CAG | Q,Q 215 | XP_005267917.1 | |
XM_011536966.2 | 813 | Intron | XP_011535268.1 | |||
XM_011536967.1 | 813 | Silent Mutation | CAA,CAG | Q,Q 215 | XP_011535269.1 |
Set Membership: |
Validated |