Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCTGAGGCCTGGCAAGACAGCCC[A/G]TCTACAGTCATTGGACATAGGGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603804 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NEURL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NEURL1 - neuralized E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH3PXD2A - SH3 and PX domains 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014631.2 | 10161 | UTR 3 | NP_055446.2 | |||
XM_005270294.4 | 10161 | UTR 3 | XP_005270351.1 | |||
XM_005270295.4 | 10161 | UTR 3 | XP_005270352.1 | |||
XM_005270297.3 | 10161 | UTR 3 | XP_005270354.1 | |||
XM_005270298.4 | 10161 | UTR 3 | XP_005270355.1 | |||
XM_006718079.3 | 10161 | UTR 3 | XP_006718142.1 | |||
XM_006718080.3 | 10161 | UTR 3 | XP_006718143.1 | |||
XM_011540385.2 | 10161 | UTR 3 | XP_011538687.1 | |||
XM_011540386.2 | 10161 | UTR 3 | XP_011538688.1 | |||
XM_011540387.2 | 10161 | UTR 3 | XP_011538689.1 | |||
XM_017016969.1 | 10161 | UTR 3 | XP_016872458.1 | |||
XM_017016970.1 | 10161 | UTR 3 | XP_016872459.1 |