Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGCAACAATCAAAAGGAATGGGG[A/G]ACTCTATGCCCAGACAGTGAGTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605080 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNGB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CNGB3 - cyclic nucleotide gated channel beta 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019098.4 | Intron | NP_061971.3 | ||||
XM_011517138.2 | Intron | XP_011515440.1 |
Set Membership: |
HapMap |