Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605363 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAD1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GAD1 - glutamate decarboxylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000817.2 | 274 | UTR 5 | NP_000808.2 | |||
NM_013445.3 | 274 | UTR 5 | NP_038473.2 | |||
XM_005246444.2 | 274 | UTR 5 | XP_005246501.1 | |||
XM_011510922.1 | 274 | Intron | XP_011509224.1 | |||
XM_017003756.1 | 274 | Intron | XP_016859245.1 | |||
XM_017003757.1 | 274 | UTR 5 | XP_016859246.1 | |||
XM_017003758.1 | 274 | UTR 5 | XP_016859247.1 |
Set Membership: |
HapMap Validated |