Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGGGTTGGCCCATAGCTATTGG[A/C]GGAAGCAAACGAAACAGAGGCATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605363 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GAD1 - glutamate decarboxylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000817.2 | Intron | NP_000808.2 | ||||
NM_013445.3 | Intron | NP_038473.2 | ||||
XM_005246444.2 | Intron | XP_005246501.1 | ||||
XM_011510922.1 | Intron | XP_011509224.1 | ||||
XM_017003756.1 | Intron | XP_016859245.1 | ||||
XM_017003757.1 | Intron | XP_016859246.1 | ||||
XM_017003758.1 | Intron | XP_016859247.1 |
Set Membership: |
Validated |