Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCGTGCGGGGCGCGACCGAGGCG[C/T]CTTTTCCTGTCCCCTGCTGTCGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300954 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CXorf40A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CXorf40A - chromosome X open reading frame 40A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171907.2 | Intron | NP_001165378.1 | ||||
NM_001171908.2 | Intron | NP_001165379.1 | ||||
NM_001171909.3 | Intron | NP_001165380.1 | ||||
NM_001324274.1 | Intron | NP_001311203.1 | ||||
NM_001324275.1 | Intron | NP_001311204.1 | ||||
NM_001324276.1 | Intron | NP_001311205.1 | ||||
NM_001324277.1 | Intron | NP_001311206.1 | ||||
NM_001324278.1 | Intron | NP_001311207.1 | ||||
NM_001324279.1 | Intron | NP_001311208.1 | ||||
NM_001324280.1 | Intron | NP_001311209.1 | ||||
NM_178124.5 | Intron | NP_835225.2 | ||||
XM_005262349.4 | Intron | XP_005262406.2 |
LINC00893 - long intergenic non-protein coding RNA 893 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101928917 - heat shock transcription factor, X-linked-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |