Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGGAGGGTGGGTGCTGGGTCCC[A/G]GGGTCCAGAAGCTGGAGAAGGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611191 MIM: 614510 MIM: 611250 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR125A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR125A - microRNA 125a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR99B - microRNA 99b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIRLET7E - microRNA let-7e | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPACA6 - sperm acrosome associated 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316972.1 | 12 | Intron | NP_001303901.1 | |||
NM_001316994.1 | 12 | Intron | NP_001303923.1 | |||
XM_017026299.1 | 12 | UTR 5 | XP_016881788.1 | |||
XM_017026300.1 | 12 | Intron | XP_016881789.1 | |||
XM_017026301.1 | 12 | Intron | XP_016881790.1 |
SPACA6P-AS - SPACA6P antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |