Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCCTGCCTCTGCACCCAGCCTG[A/T]CCACCGCCCAACAGCCTTTAGACCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608482 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100128770 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC100128770 - uncharacterized LOC100128770 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MMP25 - matrix metallopeptidase 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022468.4 | Intron | NP_071913.1 | ||||
XM_011522602.2 | Intron | XP_011520904.1 | ||||
XM_011522604.2 | Intron | XP_011520906.1 | ||||
XM_011522605.2 | Intron | XP_011520907.1 | ||||
XM_017023561.1 | Intron | XP_016879050.1 |
MMP25-AS1 - MMP25 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |