Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCAGCTGGGTGGGCCCAAGAGCA[A/G]GGCCTGTGTGGGTCCTGGTGTCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606001 MIM: 608482 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL32 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL32 - interleukin 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012631.1 | Intron | NP_001012649.1 | ||||
NM_001012632.1 | Intron | NP_001012650.1 | ||||
NM_001012633.1 | Intron | NP_001012651.1 | ||||
NM_001012634.1 | Intron | NP_001012652.1 | ||||
NM_001012635.1 | Intron | NP_001012653.1 | ||||
NM_001012636.1 | Intron | NP_001012654.1 | ||||
NM_001012718.1 | Intron | NP_001012736.1 | ||||
NM_001308078.1 | Intron | NP_001295007.1 | ||||
NM_004221.4 | Intron | NP_004212.4 |
MMP25 - matrix metallopeptidase 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MMP25-AS1 - MMP25 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |